Order Entry
Belgium
ContactUsLinkComponent
 
Ethyl cellulose, 90-110 mPa.s [5% in Toluene + Ethanol (80:20), 25 °C]
 :  TCI
 
Ethyl cellulose, 90-110 mPa.s [5% in Toluene + Ethanol (80:20), 25 °C]
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Solubility in Toluene-EtOH mix.:
    almost transparency
  • Drying loss:
    max. 2.0 %
  • Viscosity:
    90.0 to 110.0 mPa-s (5 %, Toluene:EtOH=80:20, 25 deg-C)

 

Frequently Bought Together