Order Entry
Belgium
ContactUsLinkComponent
Decyl-β-D-glucopyranoside ≥98.0% (by GC) for biochemical research
Decyl-β-D-glucopyranoside ≥98.0% (by GC) for biochemical research
Catalog # TCIAD5680-1G
Supplier:  TCI
CAS Number:  
Decyl-β-D-glucopyranoside ≥98.0% (by GC) for biochemical research
Catalog # TCIAD5680-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Water:
    max. 1.0 %

Specification Test Results

About this item

Synonyms: (2R,3R,4S,5S,6R)-2-(Decyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Decyl beta-D-glucopyranoside, Decyl β-D-glucopyranoside, Decyl glucopyranoside, DG, Decyl beta-D-glucopyranoside
 :
 :320.43 g/mol
Melting Pt:75°C
Storage Temperature:Freezer
EINECS:261-469-7
CAS Number:58846-77-8
Formula:C₁₆H₃₂O₆
MDL Number:MFCD00063297