Order Entry
Belgium
ContactUsLinkComponent
4-(3,6-Di-tert-butyl-9H-carbazol-9-yl)aniline ≥98.0% (by GC)
4-(3,6-Di-tert-butyl-9H-carbazol-9-yl)aniline ≥98.0% (by GC)
Catalog # TCIAD5592-1G
Supplier:  TCI
CAS Number:  
4-(3,6-Di-tert-butyl-9H-carbazol-9-yl)aniline ≥98.0% (by GC)
Catalog # TCIAD5592-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 96.0 %
  • Melting point:
    223.0 to 228.0 deg-C

Specification Test Results

About this item

Synonyms: 3,6-Di-tert-butyl-9-(4-aminophenyl)carbazole
 :
 :370.54 g/mol
 :GHS07
CAS Number:255829-30-2
Formula:C₂₆H₃₀N₂
MDL Number:MFCD32062891