N,4-Dimethoxy-N-methylbenzamide ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:52898-49-4
:
Formula:C₁₀H₁₃NO₃
MDL Number:MFCD02684314
:195.22 g/mol
Boiling Pt:105°C
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together