Order Entry
Belgium
ContactUsLinkComponent
Dynasore ≥98.0% (by HPLC)
Dynasore ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
Dynasore ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Green to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    8.00 to 9.00 %

Specification Test Results

Frequently Bought Together