3,6-Dibromo-9-(m-tolyl)-9H-carbazole ≥98.0% (by total nitrogen basis)
TCIAD5424-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
:
:415.13 g/mol
:GHS07
:890653-54-0
:C₁₉H₁₃Br₂N