Doripenem monohydrate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:364622-82-2
:
Formula:C₁₅H₂₄N₄O₆S₂·H₂O
MDL Number:MFCD09749887
:420.50 g/mol
Melting Pt:173°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Specific rotation [a]20/D:+32.0 to +37.0 deg (C=1, H2O)
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):12.00 to 13.50 %
Specification Test Results
Frequently Bought Together