3,9-Diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by HPLC, titration analysis)
: TCI
: 6,9-Diphenylcarbazole-3-boronic acid Pinacol ester, 3,9-Diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole, 4,4,5,5-Tetramethyl-2-(6,9-diphenylcarbazol-3-yl)-1,3,2-dioxaborolane
:1359833-28-5
:
:C₃₀H₂₈BNO₂
:MFCD29089360
:445.37 g/mol
:GHS07
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:162.0 to 166.0 deg-C
Frequently Bought Together