Order Entry
Belgium
ContactUsLinkComponent
6,9-Diphenyl-9H-carbazole-3-boronic acid (contains varying amounts of Anhydride)
6,9-Diphenyl-9H-carbazole-3-boronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
6,9-Diphenyl-9H-carbazole-3-boronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 105.0 %

Specification Test Results

Frequently Bought Together