Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Flammable
Irritant
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    69 g/l (20 °C)
  • Melting Point:
    -116.3 °C
  • Molar Mass:
    74.12 g/mol
  • Boiling Point:
    34.6 °C (1013 hPa)
  • Vapor Pressure:
    587 hPa (20 °C)
  • Flash Point:
    -40 °C
  • Explosion Limit:
    1.7 - 36 %(V)
  • Density:
    0.71 g/cm3
  • Ignition Point:
    180 °C
  • Saturation concentration:
    1776 g/m3 (20 °C) Air
  • Evaporation number:
    1

Specification Test Results

Related Information

Frequently Bought Together