Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dextrin

Dextrin

Supplier:   TCI
Dextrin
Supplier:  TCI
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,02953
  • EINECS:232-675-4
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Drying loss:
    max. 10.0 %
  • Ignition residue (Sulfate):
    max. 0.5 %
  • Reducing suger:
    max. 5.0 %
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 23,10 €
    23,10 €
    1 * 100 g
       23,10 €

  • 500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 67,10 €
    67,10 €
    1 * 500 g
       67,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.