2,7-Dibromo-9-(9-heptadecyl)carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
Synonyms: 2,7-Dibromo-9-(heptadecan-9-yl)-9H-carbazole
CAS Number:955964-73-5
:
Formula:C₂₉H₄₁Br₂N
MDL Number:MFCD16619379
:563.46 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:70.0 to 74.0 deg-C
Specification Test Results
Frequently Bought Together