3-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:960524-18-9
:
Formula:C₂₂H₃₉BO₂S
:378.42 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together