Order Entry
Belgium
ContactUsLinkComponent
 
Bianthronyl ≥97.0% (by HPLC)
 :  TCI
 
Bianthronyl ≥97.0% (by HPLC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

 

Frequently Bought Together