Chlorotrihexylsilane ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3634-67-1
:
Formula:C₁₈H₃₉ClSi
MDL Number:MFCD00000510
:319.05 g/mol
Boiling Pt:154°C
EINECS:222-851-9
UN:2987
ADR:8,II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together