

Calcifediol monohydrate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:63283-36-3
:
Formula:C₂₇H₄₄O₂·H₂O
MDL Number:MFCD08600001
:400.65 g/mol
UN:2811
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Carbon):75.50 to 80.00 %
- Water:3.0 to 5.5 %
Specification Test Results
Frequently Bought Together