- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 107.0 %
: 2-(9H-Carbazol-9-yl)benzeneboronic acid, (2-(9H-Carbazol-9-yl)phenyl)boronic acid
:
:287.13 g/mol
:GHS07
:1189047-28-6
:C₁₈H₁₄BNO₂
:MFCD30543408