4-(9-Carbazolyl)benzeneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
Synonyms: 4-(9H-9-carbozale)phenylboronic acid, 4-(9H-Carbazol-9-yl)phenylboronic acid, (4-(9H-Carbazol-9-yl)phenyl)boronic acid, 4-(9H-Carbazol-9-yl)benzeneboronic acid
CAS Number:419536-33-7
:
Formula:C₁₈H₁₄BNO₂
MDL Number:MFCD13176534
:287.125 g/mol
Melting Pt:264°C
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 107.0 %
Specification Test Results
Related Information
Frequently Bought Together