N-Boc-L-aspartic acid 4-cyclohexyl ester ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:73821-95-1
:
Formula:C₁₅H₂₅NO₆
MDL Number:MFCD00061996
:315.37 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:94.0 to 98.0 deg-C
- Specific rotation [a]20/D:-20.0 to -24.0 deg (C=2、DMF)
Specification Test Results
Frequently Bought Together