5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) ≥90.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:92557-80-7
:
Formula:C₂₅H₁₅NO₉
MDL Number:MFCD00077322
:473.39 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity (GC):min. 90.0 %
- Melting point:45.0 to 49.0 deg-C
Specification Test Results
Frequently Bought Together