Ciprofloxacin hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:86393-32-0
:
Formula:C₁₇H₁₈FN₃O₃·HCl·H₂O
MDL Number:MFCD00242856
:385.82 g/mol
Merck Index:14,02314
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Water:4.7 to 6.7 %
Specification Test Results
Frequently Bought Together