
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II)
Supplier: TCI
Certificates
About this item
CAS Number:32993-05-8
:
Formula:C₄₁H₃₅ClP₂Ru
MDL Number:MFCD00075004
:726.20 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Elemental analysis (Carbon):66.00 to 69.00 %
Specification Test Results
Frequently Bought Together