N-Carbobenzoxy-DL-norleucine ≥99.0% (by titrimetric analysis)
TCIAC0760-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Melting point:104.0 to 106.0 deg-C
: N-Cbz-DL-norleucine, Z-DL-Nle-OH
:
:265.31 g/mol
:15027-13-1
:C₁₄H₁₉NO₄
:MFCD00037271