N-Carbobenzoxy-D-phenylalanine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2448-45-5
:
Formula:C₁₇H₁₇NO₄
MDL Number:MFCD00063151
:299.33 g/mol
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 97.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:85.0 to 89.0 deg-C
Specification Test Results
Frequently Bought Together