
N-Carbobenzoxy-L-aspartic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1152-61-0
:
Formula:C₁₂H₁₃NO₆
MDL Number:MFCD00002719
:267.24 g/mol
EINECS:214-568-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:117.0 to 122.0 deg-C
- Specific rotation [a]20/D:+9.0 to +10.0 deg (C=7, AcOH)
Specification Test Results
Frequently Bought Together