Copper PAN
Supplier: TCI
About this item
Synonyms: Bis[1-(2-pyridylazo)-2-naphtholato]copper(II)
- CAS Number:23299-85-6
- MDL Number:MFCD00067176
2 Options Available Below
Product Details & Documents
Synonyms: Bis[1-(2-pyridylazo)-2-naphtholato]copper(II)
- CAS Number:23299-85-6
- MDL Number:MFCD00067176
Specifications for Product Family
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Absorbance:0.500 to 0.550 (50 %, Dioxane, 466.0 to 472.0 nm)
Product Family Options
Product Information
- PkAvailabilityPrice



