N-Chloroacetyl-DL-norleucine ≥99.0% (by titrimetric analysis)
TCIAC0102-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Melting point:112.0 to 114.0 deg-C
- Solubility in Methanol:almost transparency
:
:207.65 g/mol
:GHS07
:67206-26-2
:C₈H₁₄ClNO₃
:MFCD00059650