N-Carbamoyl-DL-aspartic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0029-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
:
:176.13 g/mol
Storage Temperature:Refrigerator
EINECS:213-096-6
CAS Number:923-37-5
Formula:C₅H₈N₂O₅
MDL Number:MFCD00042822