Biotin-PEG2-NHS
Catalog # TCIAB6097-1SET
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Functionality test:to pass test
Specification Test Results
About this item
:
:500.57 g/mol
Storage Temperature:Freezer
CAS Number:596820-83-6
:MFCD11974976
Formula:C₂₁H₃₂N₄O₈S
MDL Number:MFCD00467154