Biotin-HPDP
Supplier: TCI
About this item
Synonyms: N-[6-(Biotinamido)hexyl]-3'-(2-pyridyldithio)propionamide, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-N-(6-(3-(pyridin-2-yldisulphanyl)propanamido)hexyl)pentanamide
- CAS Number:129179-83-5
- Formula:C₂₄H₃₇N₅O₃S₃
- MDL Number:MFCD00078541
- Molecular Weight:539.772 g/mol
- Storage Temperature:Freezer
2 Options Available Below
Product Details & Documents
Synonyms: N-[6-(Biotinamido)hexyl]-3'-(2-pyridyldithio)propionamide, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-N-(6-(3-(pyridin-2-yldisulphanyl)propanamido)hexyl)pentanamide
- CAS Number:129179-83-5
- Formula:C₂₄H₃₇N₅O₃S₃
- MDL Number:MFCD00078541
- Molecular Weight:539.772 g/mol
- Storage Temperature:Freezer
Specifications for Product Family
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):12.30 to 13.60 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



