Biotin-PEG6-Maleimide ≥96.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1808990-66-0
:
Formula:C₃₁H₅₁N₅O₁₁S
:701.83 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 96.0 area%
- Elemental analysis (Nitrogen):9.50 to 10.50 %
Specification Test Results
Frequently Bought Together