
1-Butyl-1-methylpyrrolidinium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
TCIAB5398-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Ion exchange titration):min. 98.0 %
:
:322.39 g/mol
:-19°C
:GHS07
:1057745-51-3
:C₉H₂₀F₂N₂O₄S₂
:MFCD30749250
:–19°C