9-(4-Biphenylyl)-3-(4-bromophenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
Synonyms: 9-([1,1'-Biphenyl]-4-yl)-3-(4-bromophenyl)-9H-carbazole
CAS Number:1028648-25-0
:
Formula:C₃₀H₂₀BrN
MDL Number:MFCD22571689
:474.40 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:192.0 to 196.0 deg-C
Specification Test Results
Frequently Bought Together