Order Entry
Belgium
ContactUsLinkComponent
 
9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)
 :  TCI
 
undefined
9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 96.0 %
  • Melting point:
    237.0 to 241.0 deg-C

 

Frequently Bought Together