
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:261904-39-6
:
Formula:C₄H₇BrO₃
MDL Number:MFCD09751022
:183.0006 g/mol
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:113.0 to 117.0 deg-C
- Specific rotation [a]20/D:+10.0 to +12.0 deg (C=2.6, MeOH)
Specification Test Results
Frequently Bought Together