tert-Butylamine hydroiodide ≥97.0% (by total nitrogen and titration analysis)
: TCI
:39557-45-4
:
:C₄H₁₁N
:201.05 g/mol
:GHS07
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Frequently Bought Together