Order Entry
Belgium
ContactUsLinkComponent
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine ≥98.0% (by HPLC, titration analysis)
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB4184-5G
Supplier:  TCI
CAS Number:  
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB4184-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -8.0 to -10.0 deg (C=1, DMF)

Specification Test Results

About this item

 :
 :454.52 g/mol
Storage Temperature:Refrigerator
CAS Number:109425-55-0
Formula:C₂₅H₃₀N₂O₆
MDL Number:MFCD00065668