1-Benzhydrylazetidine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 1-(Diphenylmethyl)azetidine-3-carboxylic Acid
CAS Number:36476-87-6
:MFCD01566539
Formula:C₁₇H₁₇NO₂
MDL Number:MFCD01566538
:267.33 g/mol
Melting Pt:198°C
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together