
4-Bromo-3-hydroxy-2-naphthoic acid ≥97.0% (by GC, titration analysis)
: TCI
:2208-15-3
:
:C₁₁H₇BrO₃
:MFCD00094166
:267.08 g/mol
:231°C
:218-632-2
:GHS07
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Frequently Bought Together