7-Bromo-3-hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1779-11-9
:
Formula:C₁₁H₇BrO₃
MDL Number:MFCD00137823
:267.08 g/mol
Melting Pt:262°C
:GHS07
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together