N-(tert-Butoxycarbonyl)-D-aspartic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2965-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+3.0 to +7.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
:233.22 g/mol
CAS Number:62396-48-9
Formula:C₉H₁₅NO₆
MDL Number:MFCD00798618