1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:30925-18-9
:
Formula:C₁₆H₂₁NO₆
MDL Number:MFCD00065563
:323.35 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:97.0 to 101.0 deg-C
- Specific rotation [a]20/D:-21.0 to -26.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together