Order Entry
Belgium
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-L-aspartic acid ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-L-aspartic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2270-5G
Supplier:  TCI
CAS Number:  
N-(tert-Butoxycarbonyl)-L-aspartic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB2270-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -3.0 to -7.0 deg (C=1, MeOH)

Specification Test Results

About this item

 :
 :233.22 g/mol
EINECS:237-294-7
CAS Number:13726-67-5
Formula:C₉H₁₅NO₆
MDL Number:MFCD00037279