Order Entry
Belgium
ContactUsLinkComponent
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    92.0 to 95.0 deg-C
  • Specific rotation [a]20/D:
    -5.0 to -1.0 deg (C=1, AcOH)

Specification Test Results

Related Information

Frequently Bought Together