
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:59831-02-6
:
Formula:C₂₇H₂₆Cl₂P₂Pd
MDL Number:MFCD03844773
:589.77 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together