Order Entry
Belgium
ContactUsLinkComponent
Bis(tricyclohexylphosphine)palladium(II) dichloride ≥97.0% (by titrimetric analysis)
Bis(tricyclohexylphosphine)palladium(II) dichloride ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Bis(tricyclohexylphosphine)palladium(II) dichloride ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Purity (Chelometric Titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together