Methyl decanoate, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:110-42-9
:
Formula:C₁₁H₂₂O₂
MDL Number:MFCD00009580
:186.29 g/mol
Boiling Pt:224 °C (1013 hPa)
Melting Pt:–18 °C
Density:0.8724 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:94 °C
EINECS:203-766-6
Frequently Bought Together