Order Entry
Belgium
ContactUsLinkComponent
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Irritant
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together