
Bis(acetonitrile)palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14592-56-4
:
Formula:C₄H₆Cl₂N₂Pd
MDL Number:MFCD00013122
:259.43 g/mol
EINECS:238-637-3
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light yellow to Brown powder to crystaline
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together