4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7536-58-5
:
Formula:C₁₆H₂₁NO₆
MDL Number:MFCD00065564
:323.35 g/mol
EINECS:231-406-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:102.0 to 104.0 deg-C
- Specific rotation [a]20/D:-19.0 to -21.0 deg (C=2, DMF)
- Solubility in N,N-DMF:almost transparency
Specification Test Results
Frequently Bought Together